Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam81b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760605.1 | XP_008758827.1 | 3 - 4 | 389 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |