Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pm20d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109068.1 | NP_001102538.1 | 10 - 11 | 1165 | 64 | |
GenBank mRNA | FQ210945.1 | - | 6 - 7 | 452 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |