Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079689.2 | NP_001073157.1 | 8 - 9 | 1316 | 129 | |
NM_001270846.1 | NP_001257775.1 | 9 - 10 | 965 | 129 | ||
NM_001270847.1 | NP_001257776.1 | 8 - 9 | 864 | 129 | ||
NM_001270848.1 | NP_001257777.1 | 8 - 9 | 949 | 129 | ||
XM_008769542.1 | XP_008767764.1 | 7 - 8 | 1245 | 129 | ||
GenBank mRNA | DQ286232.1 | - | 7 - 8 | 842 | 129 | |
DQ923316.1 | - | 7 - 8 | 787 | 129 | ||
DQ923317.1 | - | 8 - 9 | 949 | 129 | ||
DQ923318.1 | - | 8 - 9 | 865 | 129 | ||
DQ923319.1 | - | 9 - 10 | 965 | 129 | ||
DQ923320.1 | - | 8 - 9 | 1307 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |