Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zp3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270845.1 | NP_001257774.1 | 1 - 2 | 72 | 71 | |
NM_182815.2 | NP_877967.2 | 1 - 2 | 72 | 71 | ||
GenBank mRNA | AY278363.1 | - | 1 - 2 | 72 | 71 | |
AY278364.1 | - | 1 - 2 | 72 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |