Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191929.1 | NP_001178858.1 | 2 - 3 | 209 | 76 | |
XM_006250187.2 | XP_006250249.1 | 4 - 5 | 409 | 76 | ||
XM_006250188.2 | XP_006250250.1 | 4 - 5 | 405 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Rattus norvegicus Probe spans exons |