Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007759.1 | NP_001007760.1 | 1 - 2 | 183 | 101 | |
GenBank mRNA | BC083738.1 | - | 1 - 2 | 183 | 7191669 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |