Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173133.2 | NP_775156.2 | 2 - 3 | 708 | 90 | |
GenBank mRNA | BC086410.1 | - | 2 - 3 | 678 | 90 | |
BC097932.1 | - | 2 - 3 | 684 | 7386998 | 90 | |
FQ226770.1 | - | 1 - 2 | 256 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |