Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173133.2 | NP_775156.2 | 1 - 2 | 119 | 141 | |
GenBank mRNA | X67877.1 | - | 1 - 2 | 119 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |