Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncaip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107379.1 | NP_001100849.1 | 2 - 3 | 428 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |