Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254761.2 | XP_006254823.1 | 6 - 7 | 726 | 102 | |
GenBank mRNA | X14254.1 | - | 6 - 7 | 632 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |