Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019314.1 | NP_062187.1 | 2 - 3 | 582 | 130 | |
XM_006254682.2 | XP_006254744.1 | 2 - 3 | 2135 | 130 | ||
XM_006254683.2 | XP_006254745.1 | 3 - 4 | 3208 | 130 | ||
GenBank mRNA | U69882.1 | - | 2 - 3 | 582 | 130 |
Gene Symbol: LOC103690147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772181.1 | XP_008770403.1 | 2 - 3 | 2137 | 130 | |
XM_008772182.1 | XP_008770404.1 | 3 - 4 | 3166 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Rattus norvegicus Probe spans exons |