Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019314.1 | NP_062187.1 | 5 - 6 | 1254 | 100 | |
XM_006254682.2 | XP_006254744.1 | 5 - 6 | 2807 | 100 | ||
XM_006254683.2 | XP_006254745.1 | 6 - 7 | 3880 | 100 | ||
GenBank mRNA | JN857942.1 | - | 5 - 6 | 1254 | 100 | |
U69882.1 | - | 5 - 6 | 1254 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |