Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033887.1 | NP_001029059.1 | 6 - 7 | 1062 | 62 | |
XM_006254746.2 | XP_006254808.1 | 7 - 8 | 1258 | 62 | ||
GenBank mRNA | BC101903.1 | - | 6 - 7 | 1062 | 7934311 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |