Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052809.1 | NP_434696.1 | 3 - 4 | 593 | 107 | |
GenBank mRNA | BC070509.1 | - | 3 - 4 | 584 | 7100321 | 107 |
FQ218376.1 | - | 2 - 3 | 230 | 107 | ||
FQ219282.1 | - | 2 - 3 | 230 | 107 | ||
M35266.1 | - | 3 - 4 | 593 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |