Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237372.2 | XP_006237434.1 | 9 - 10 | 1641 | 138 | |
XM_006237373.2 | XP_006237435.1 | 9 - 10 | 1638 | 138 | ||
XM_006237374.2 | XP_006237436.1 | 9 - 10 | 1639 | 138 | ||
XM_006237375.2 | XP_006237437.1 | 8 - 9 | 1632 | 138 | ||
XM_006237376.2 | XP_006237438.1 | 8 - 9 | 1629 | 138 | ||
XM_008763292.1 | XP_008761514.1 | 9 - 10 | 1666 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |