Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107889.1 | NP_001101359.1 | 4 - 5 | 506 | 77 | |
XM_006237363.2 | XP_006237425.1 | 5 - 6 | 496 | 77 | ||
XM_006237364.2 | XP_006237426.1 | 5 - 6 | 496 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |