Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017253.2 | NP_058949.1 | 6 - 7 | 845 | 56 | |
XM_008763370.1 | XP_008761592.1 | 6 - 7 | 858 | 56 | ||
XM_008763371.1 | XP_008761593.1 | 6 - 7 | 1108 | 56 | ||
XM_008763372.1 | XP_008761594.1 | 7 - 8 | 934 | 56 | ||
XM_008763373.1 | XP_008761595.1 | 6 - 7 | 1037 | 56 | ||
GenBank mRNA | AF165887.1 | - | 4 - 5 | 602 | 56 | |
AJ278701.1 | - | 6 - 7 | 839 | 56 | ||
BC087710.1 | - | 6 - 7 | 845 | 7315507 | 56 | |
U35774.1 | - | 6 - 7 | 811 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |