Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031633.3 | NP_113821.2 | 8 - 9 | 1206 | 106 | |
XM_006237416.2 | XP_006237478.1 | 6 - 7 | 1149 | 106 | ||
XM_006237417.2 | XP_006237479.1 | 6 - 7 | 1146 | 106 | ||
XM_006237418.2 | XP_006237480.1 | 7 - 8 | 1173 | 106 | ||
XM_006237419.2 | XP_006237481.1 | 5 - 6 | 1101 | 106 | ||
XM_006237420.2 | XP_006237482.1 | 6 - 7 | 1125 | 106 | ||
XM_006237421.2 | XP_006237483.1 | 7 - 8 | 1170 | 106 | ||
GenBank mRNA | U83112.1 | - | 8 - 9 | 1206 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |