Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031548.2 | NP_113736.1 | 4 - 5 | 1434 | 153 | |
GenBank mRNA | BC070902.1 | - | 4 - 5 | 1434 | 7100777 | 153 |
EU627515.1 | - | 4 - 5 | 1069 | 153 | ||
U54699.1 | - | 4 - 5 | 1101 | 153 | ||
U54700.1 | - | 4 - 5 | 1101 | 153 | ||
X70497.1 | - | 4 - 5 | 1147 | 153 | ||
X70521.1 | - | 4 - 5 | 1122 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |