Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232135.2 | XP_006232197.1 | 2 - 3 | 677 | 99 | |
GenBank mRNA | AF068746.2 | - | 1 - 2 | 376 | 99 | |
AF305780.1 | - | 2 - 3 | 650 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |