Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012533.1 | NP_036665.1 | 2 - 3 | 143 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |