Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mynn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012178.1 | NP_001012178.1 | 2 - 3 | 209 | 68 | |
XM_006232234.2 | XP_006232296.1 | 2 - 3 | 1123 | 68 | ||
GenBank mRNA | BC089853.1 | - | 2 - 3 | 209 | 7379696 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |