Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044696.1 | NP_001038161.1 | 4 - 5 | 572 | 83 | |
XM_006232235.2 | XP_006232297.1 | 5 - 6 | 543 | 83 | ||
XM_008760937.1 | XP_008759159.1 | 5 - 6 | 634 | 83 | ||
XM_008760938.1 | XP_008759160.1 | 4 - 5 | 330 | 83 | ||
XM_008760939.1 | XP_008759161.1 | 4 - 5 | 526 | 83 | ||
GenBank mRNA | BC097432.1 | - | 4 - 5 | 572 | 7455342 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |