Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191702.1 | NP_001178631.1 | 4 - 5 | 3685 | 91 | |
XM_006232162.2 | XP_006232224.1 | 4 - 5 | 3791 | 91 | ||
XM_006232164.2 | XP_006232226.1 | 4 - 5 | 3656 | 91 | ||
XM_008760867.1 | XP_008759089.1 | 4 - 5 | 4151 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |