Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232237.2 | XP_006232299.1 | 6 - 7 | 2941 | 74 | |
XM_008760948.1 | XP_008759170.1 | 5 - 6 | 3947 | 74 | ||
XM_008760949.1 | XP_008759171.1 | 5 - 6 | 3807 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |