Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106426.1 | NP_001099896.1 | 2 - 3 | 284 | 113 | |
GenBank mRNA | FQ224246.1 | - | 2 - 3 | 235 | 113 | |
FQ224437.1 | - | 2 - 3 | 237 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |