Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrriq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760970.1 | XP_008759192.1 | 2 - 3 | 460 | 65 | |
GenBank mRNA | BC168195.1 | - | 1 - 2 | 310 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |