Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053702.3 | NP_446154.3 | 5 - 6 | 1059 | 73 | |
XM_006232266.2 | XP_006232328.1 | 6 - 7 | 1196 | 73 | ||
XM_008760903.1 | XP_008759125.1 | 6 - 7 | 1191 | 73 | ||
GenBank mRNA | AY623027.1 | - | 4 - 5 | 1033 | 73 | |
AY623028.1 | - | 5 - 6 | 1438 | 73 | ||
BC127462.1 | - | 6 - 7 | 1152 | 8373429 | 73 | |
BF406951.1 | - | 2 - 3 | 234 | 73 | ||
CV114678.1 | - | 2 - 3 | 326 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |