Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760719.1 | XP_008758941.1 | 4 - 5 | 743 | 57 | |
GenBank mRNA | D78197.1 | - | 4 - 5 | 555 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |