Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130747.1 | NP_570103.1 | 8 - 9 | 945 | 82 | |
XM_008760628.1 | XP_008758850.1 | 9 - 10 | 1440 | 82 | ||
GenBank mRNA | AB040609.1 | - | 8 - 9 | 945 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |