Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107650.2 | NP_001101120.2 | 10 - 11 | 1099 | 79 | |
NR_073139.1 | 10 - 11 | 952 | 79 | |||
NR_073140.1 | 9 - 10 | 899 | 79 | |||
XM_008760726.1 | XP_008758948.1 | 10 - 11 | 1161 | 79 | ||
XM_008760728.1 | XP_008758950.1 | 9 - 10 | 1009 | 79 | ||
GenBank mRNA | BC089220.1 | - | 10 - 11 | 916 | 79 | |
BC161924.1 | - | 9 - 10 | 876 | 8370925 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |