Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107650.2 | NP_001101120.2 | 7 - 8 | 677 | 91 | |
NR_073139.1 | 7 - 8 | 628 | 91 | |||
NR_073140.1 | 6 - 7 | 575 | 91 | |||
XM_008760726.1 | XP_008758948.1 | 7 - 8 | 739 | 91 | ||
XM_008760727.1 | XP_008758949.1 | 7 - 8 | 742 | 91 | ||
XM_008760728.1 | XP_008758950.1 | 6 - 7 | 587 | 91 | ||
GenBank mRNA | BC089220.1 | - | 7 - 8 | 592 | 91 | |
BC161924.1 | - | 6 - 7 | 552 | 8370925 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |