Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138516.1 | NP_612525.1 | 12 - 13 | 1728 | 92 | |
GenBank mRNA | BC166778.1 | - | 12 - 13 | 1728 | 7934797 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |