Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013029.1 | NP_037161.1 | 3 - 4 | 905 | 138 | |
GenBank mRNA | U55938.1 | - | 3 - 4 | 905 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |