Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012181.2 | NP_001012181.1 | 4 - 5 | 1174 | 126 | |
NM_001033909.1 | NP_001029081.1 | 6 - 7 | 854 | 126 | ||
XM_006232325.2 | XP_006232387.1 | 7 - 8 | 1056 | 126 | ||
XM_006232327.2 | XP_006232389.1 | 4 - 5 | 3242 | 126 | ||
XM_006232328.1 | XP_006232390.1 | 5 - 6 | 568 | 126 | ||
XM_006232329.1 | XP_006232391.1 | 5 - 6 | 532 | 126 | ||
XM_008760992.1 | XP_008759214.1 | 6 - 7 | 1439 | 126 | ||
GenBank mRNA | BC083585.1 | - | 7 - 8 | 1006 | 7131145 | 126 |
BC083598.1 | - | 4 - 5 | 526 | 7132941 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |