Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017020.3 | NP_058716.2 | 4 - 5 | 655 | 69 | |
XM_006232587.2 | XP_006232649.1 | 4 - 5 | 2089 | 69 | ||
XM_006232588.2 | XP_006232650.1 | 4 - 5 | 2088 | 69 | ||
XM_008761127.1 | XP_008759349.1 | 4 - 5 | 2084 | 69 | ||
GenBank mRNA | M58587.1 | - | 4 - 5 | 744 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |