Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spaca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105820.1 | NP_001099290.1 | 4 - 5 | 676 | 70 | |
XM_006246983.2 | XP_006247045.1 | 3 - 4 | 571 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |