Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spaca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105820.1 | NP_001099290.1 | 5 - 6 | 755 | 113 | |
XM_006246983.2 | XP_006247045.1 | 4 - 5 | 650 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |