Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itfg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009701.1 | NP_001009701.1 | 1580 | 57 | ||
GenBank mRNA | BC088404.1 | - | 1580 | 7108395 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |