Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itfg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009701.1 | NP_001009701.1 | 1476 | 78 | ||
GenBank mRNA | BC088404.1 | - | 1476 | 7108395 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |