Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031638.1 | NP_001026808.1 | 2 - 3 | 537 | 92 | |
GenBank mRNA | BC098931.1 | - | 2 - 3 | 537 | 7368498 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |