Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC061984.1 | - | 1 - 2 | 336 | 5599180 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |