Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025664.1 | NP_001020835.2 | 1 - 2 | 568 | 94 | |
NM_001042561.1 | NP_001036026.1 | 6 - 7 | 980 | 94 | ||
XM_008768016.1 | XP_008766238.1 | 6 - 7 | 1903 | 94 | ||
GenBank mRNA | BC098720.1 | - | 1 - 2 | 568 | 94 | |
BC167746.1 | - | 6 - 7 | 983 | 8364583 | 94 | |
CK473503.1 | - | 4 - 5 | 463 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |