Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105774.1 | NP_001099244.1 | 1 - 2 | 131 | 56 | |
GenBank mRNA | FQ213277.1 | - | 1 - 2 | 52 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |