Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105774.1 | NP_001099244.1 | 3 - 4 | 445 | 121 | |
XM_006245937.2 | XP_006245999.1 | 3 - 4 | 500 | 121 | ||
GenBank mRNA | BC161966.1 | - | 3 - 4 | 443 | 9028415 | 121 |
FQ213277.1 | - | 3 - 4 | 366 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |