Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031972.1 | NP_114178.1 | 8 - 9 | 1285 | 68 | |
GenBank mRNA | BC070924.1 | - | 8 - 9 | 1318 | 7096634 | 68 |
J03637.1 | - | 8 - 9 | 1285 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |