Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013965.1 | NP_001013987.1 | 2 - 3 | 712 | 112 | |
GenBank mRNA | AB194013.1 | - | 2 - 3 | 597 | 112 | |
BC079304.1 | - | 2 - 3 | 712 | 7134598 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |