Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153476.1 | NP_703206.1 | 6 - 7 | 1385 | 107 | |
GenBank mRNA | AY128946.1 | - | 6 - 7 | 1385 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |