Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247437.1 | XP_006247499.1 | 8 - 9 | 1752 | 57 | |
XM_006247438.1 | XP_006247500.1 | 8 - 9 | 1752 | 57 | ||
XM_008768198.1 | XP_008766420.1 | 6 - 7 | 2056 | 57 | ||
XM_008768199.1 | XP_008766421.1 | 7 - 8 | 2212 | 57 | ||
XM_008768200.1 | XP_008766422.1 | 6 - 7 | 3207 | 57 | ||
XM_220918.8 | XP_220918.6 | 10 - 11 | 2056 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |