Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011894.1 | NP_001011894.1 | 6 - 7 | 602 | 62 | |
GenBank mRNA | BC089889.1 | - | 6 - 7 | 602 | 7374359 | 62 |
FQ232375.1 | - | 4 - 5 | 287 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |